Anti-Secretory component (free and bound) Mouse Monoclonal Antibody [clone: NI 194-4 (A89-039)]

Supplier: Abnova
MAB10075
10681-966EA 530.63 USD
10681-966
Anti-Secretory component (free and bound) Mouse Monoclonal Antibody [clone: NI 194-4 (A89-039)]
Antibodies
Mouse monoclonal antibody raised against secretory component (free and bound).

Recommended Dilutions: ELISA: 1:500; Western Blot: 1:1000; Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: Secretory component (free and bound)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: NI 194-4 (A89-039)
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR