Anti-NOS3 Mouse Monoclonal Antibody [clone: M232]

Supplier: Abnova
MAB1391
10675-064EA 428 USD
10675-064
Anti-NOS3 Mouse Monoclonal Antibody [clone: M232]
Antibodies
Mouse monoclonal antibody raised against synthetic phosphopeptide of Nos3.

Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:500

Type: Primary
Antigen: NOS3
Clonality: Monoclonal
Clone: M232
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR