Anti-CEA Mouse Monoclonal Antibody

Supplier: Abnova
MAB1393
10626-940EA 329 USD
10626-940
Anti-CEA Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against native Carcino Embryonic Antigen.

Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR