Anti-FSHB Mouse Monoclonal Antibody

Supplier: Abnova
MAB1408
10627-458EA 329 USD
10627-458
Anti-FSHB Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against native FSHB.

Type: Primary
Antigen: FSHB
Clonality: Unconjugated
Clone: Monoclonal
Conjugation:
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR