Anti-CALD1 Mouse Monoclonal Antibody [clone: CD-7]

Supplier: Abnova
MAB1430
10705-086EA 428.44 USD
10705-086
Anti-CALD1 Mouse Monoclonal Antibody [clone: CD-7]
Antibodies
Mouse monoclonal antibody raised against native Caldesmon.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml

Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: CD-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Turkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR