Anti-CD4 Mouse Monoclonal Antibody [clone: CA-4]

Supplier: Abnova
MAB1434
10706-462EA 428.44 USD
10706-462
Anti-CD4 Mouse Monoclonal Antibody [clone: CA-4]
Antibodies
Mouse monoclonal antibody raised against recombinant CD4.

Recommended Dilutions: Immunohistochemistry (Frozen sections): 1 ?g/ml

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: CA-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR