Anti-Adenoviruses (type 1) Mouse Monoclonal Antibody [clone: 8C4]

Supplier: Abnova
MAB10114
10688-214EA 345.64 USD
10688-214
Anti-Adenoviruses (type 1) Mouse Monoclonal Antibody [clone: 8C4]
Antibodies
Mouse monoclonal antibody raised against native Adenoviruses (type 1).

Type: Primary
Antigen: Adenoviruses (type 1)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 8C4
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Bovine, Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR