Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]

Supplier: Abnova
MAB1437
10707-168EA 427.45 USD
10707-168
Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CDC6.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CDC6
Clonality: Monoclonal
Clone: IMD-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR