Anti-CK7 Mouse Monoclonal Antibody [clone: CK-7]

Supplier: Abnova
MAB1442
10722-980EA 428.44 USD
10722-980
Anti-CK7 Mouse Monoclonal Antibody [clone: CK-7]
Antibodies
Mouse monoclonal antibody raised against native KRT7.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml; Immunohistochemistry (Frozen sections): 2-4 ?g/ml

Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone: CK-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR