Anti-GAP43 Mouse Monoclonal Antibody [clone: GAP-8A12]

Supplier: Abnova
MAB1455
10715-582EA 428.44 USD
10715-582
Anti-GAP43 Mouse Monoclonal Antibody [clone: GAP-8A12]
Antibodies
Mouse monoclonal antibody raised against native Gap43.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml; Immunohistochemistry (Frozen sections): 1-2 ?g/ml

Type: Primary
Antigen: Gap43
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: GAP-8A12
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Chicken, Human, Mouse, Rat, Snake
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR