Anti-MAPK1 Mouse Monoclonal Antibody [clone: IL-13]

Supplier: Abnova
MAB1463
10674-898EA 428 USD
10674-898
Anti-MAPK1 Mouse Monoclonal Antibody [clone: IL-13]
Antibodies
Mouse monoclonal antibody raised against synthetic phosphopeptide of MAPK1.

Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 0.4-1 ?g/ml

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: IL-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR