Anti-MUC5AC Mouse Monoclonal Antibody [clone: MG-31]

Supplier: Abnova
MAB1466
10726-242EA 428.44 USD
10726-242
Anti-MUC5AC Mouse Monoclonal Antibody [clone: MG-31]
Antibodies
Mouse monoclonal antibody raised against native MUC5AC.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml

Type: Primary
Antigen: MUC5AC
Clonality: Monoclonal
Clone: MG-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR