Anti-CDKN2A Mouse Monoclonal Antibody [clone: IMD-16]

Supplier: Abnova
MAB1472
10707-392EA 428.44 USD
10707-392
Anti-CDKN2A Mouse Monoclonal Antibody [clone: IMD-16]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CDKN2A.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry (Frozen sections): 1-2 ?g/ml

Type: Primary
Antigen: CDKN2A
Clonality: Monoclonal
Clone: IMD-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR