Anti-Pan-Cadherin Mouse Monoclonal Antibody [clone: PC-79]

Supplier: Abnova
MAB1477
10728-410EA 428.44 USD
10728-410
Anti-Pan-Cadherin Mouse Monoclonal Antibody [clone: PC-79]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of cadherin.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml

Type: Primary
Antigen: Pan-Cadherin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: PC-79
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Chicken, Human, Mouse, Rabbit, Rat, Snake
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR