Anti-HA (H5N1) Mouse Monoclonal Antibody [clone: IT-1056]

Supplier: Abnova
MAB10132
10720-758EA 530.63 USD
10720-758
Anti-HA (H5N1) Mouse Monoclonal Antibody [clone: IT-1056]
Antibodies
Mouse monoclonal antibody raised against partial recombinant HA (H5N1).

Type: Primary
Antigen: HA (H5N1)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: IT-1056
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR