Anti-DYKDDDDK tag Monoclonal Antibody [clone: 100fg]

Supplier: Abnova
MAB10142
10711-426EA 329 USD
10711-426
Anti-DYKDDDDK tag Monoclonal Antibody [clone: 100fg]
Antibodies
Mouse monoclonal antibody raised against DYKDDDDK tag.

Type: Primary
Antigen: DYKDDDDK tag
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 100fg
Epitope:
Host: IgG1 kappa
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR