Anti-GATA3 Rat Monoclonal Antibody [clone: KT63]

Supplier: Abnova
MAB10148
10715-634EA 346 USD
10715-634
Anti-GATA3 Rat Monoclonal Antibody [clone: KT63]
Antibodies
Rat monoclonal antibody raised against Gata3.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml

Type: Primary
Antigen: Gata3
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: KT63
Epitope:
Host: IgG2a
Isotype: Rat
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR