Anti-IgG1 Monoclonal Antibody (PE (Phycoerythrin)) [clone: MOPC-21]

Supplier: Abnova
MAB10156
10673-982EA 329 USD
10673-982
Anti-IgG1 Monoclonal Antibody (PE (Phycoerythrin)) [clone: MOPC-21]
Antibodies
Mouse monoclonal antibody was chosen by screening on variety of resting, activated, live and fixed rat and human tissues.

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)
Epitope:
Host:
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR