Anti-AFP Mouse Monoclonal Antibody [clone: AP-1]

Supplier: Abnova
MAB1508
10688-376EA 428.44 USD
10688-376
Anti-AFP Mouse Monoclonal Antibody [clone: AP-1]
Antibodies
Mouse monoclonal antibody raised against native AFP.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: AP-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR