Anti-MAP2 Mouse Monoclonal Antibody [clone: MP-2]

Supplier: Abnova
MAB1521
10724-638EA 428.44 USD
10724-638
Anti-MAP2 Mouse Monoclonal Antibody [clone: MP-2]
Antibodies
Mouse monoclonal antibody raised against native Map2.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: MP-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR