Anti-PCNA Mouse Monoclonal Antibody [clone: IML-83]

Supplier: Abnova
MAB1524
10733-392EA 428.44 USD
10733-392
Anti-PCNA Mouse Monoclonal Antibody [clone: IML-83]
Antibodies
Mouse monoclonal antibody raised against native PCNA.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 0.4-1 ?g/ml; Immunohistochemistry (Frozen sections): 0.4-1 ?g/ml

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: IML-83
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR