Anti-CD19 Mouse Monoclonal Antibody [clone: 4G7]

Supplier: Abnova
MAB10163
10706-028EA 329 USD
10706-028
Anti-CD19 Mouse Monoclonal Antibody [clone: 4G7]
Antibodies
Mouse monoclonal antibody raised against CD19.

Recommended Dilutions: Flow Cytometry: 1 ?g/ml

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 4G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR