Anti-FCGR3A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3G8]

Supplier: Abnova
MAB10168
10714-564EA 329 USD
10714-564
Anti-FCGR3A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3G8]
Antibodies
Mouse monoclonal antibody raised against FCGR3A.

Recommended Dilutions: Flow Cytometry: 6 ?g/ml

Type: Primary
Antigen: FCGR3A
Clonality: PE (Phycoerythrin)
Clone: Monoclonal
Conjugation: 3G8
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human, Non-Human Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR