Anti-SELP Mouse Monoclonal Antibody [clone: AK4]

Supplier: Abnova
MAB10170
10737-502EA 329 USD
10737-502
Anti-SELP Mouse Monoclonal Antibody [clone: AK4]
Antibodies
Mouse monoclonal antibody raised against SELP.

Recommended Dilutions: Flow Cytometry: 1.5 ?g/ml

Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: AK4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-human Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR