Anti-GRAP2 Mouse Monoclonal Antibody [clone: UW40]

Supplier: Abnova
MAB10180
10716-372EA 329 USD
10716-372
Anti-GRAP2 Mouse Monoclonal Antibody [clone: UW40]
Antibodies
Mouse monoclonal antibody raised against recombinant GRAP2.

Recommended Dilutions: Flow Cytometry: 1 ?g/ml

Type: Primary
Antigen: GRAP2
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: UW40
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR