Anti-SMC3 Rat Monoclonal Antibody [clone: KT75]

Supplier: Abnova
MAB10224
10738-120EA 346 USD
10738-120
Anti-SMC3 Rat Monoclonal Antibody [clone: KT75]
Antibodies
Rat monoclonal antibody raised against recombinant Smc3.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: SMC3
Clonality: Monoclonal
Clone: KT75
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR