Anti-LAMP2 Mouse Monoclonal Antibody [clone: H4B4]

Supplier: Abnova
MAB10238
10723-250EA 329 USD
10723-250
Anti-LAMP2 Mouse Monoclonal Antibody [clone: H4B4]
Antibodies
Mouse monoclonal antibody raised against of LAMP2.

Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone: H4B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR