Anti-TP53 Rabbit Monoclonal Antibody [clone: S11-K]

Supplier: Abnova
MAB1626
10692-178EA 345.64 USD
10692-178
Anti-TP53 Rabbit Monoclonal Antibody [clone: S11-K]
Antibodies
Rabbit monoclonal antibody raised against synthetic peptide of TP53.

Recommended Dilutions: Western Blot: 1:2000; ELISA: 1:10000-1:20000

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: S11-K
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR