Anti-NOS2 Rabbit Monoclonal Antibody [clone: K13-A]

Supplier: Abnova
MAB1627
10727-418EA 346 USD
10727-418
Anti-NOS2 Rabbit Monoclonal Antibody [clone: K13-A]
Antibodies
Rabbit monoclonal antibody raised against synthetic peptide of NOS2.

Recommended Dilutions: Western Blot: 1:2000; ELISA: 1:20000-1:50000

Type: Primary
Antigen: NOS2
Clonality: Monoclonal
Clone: K13-A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR