Anti-PRKCB Rabbit Monoclonal Antibody [clone: A10-F]

Supplier: Abnova
MAB1630
10692-056EA 346 USD
10692-056
Anti-PRKCB Rabbit Monoclonal Antibody [clone: A10-F]
Antibodies
Rabbit monoclonal antibody raised against synthetic peptide of PKC-beta 1.

Recommended Dilutions: Western Blot: 1:5000; ELISA: 1:20000-1:50000

Type: Primary
Antigen: PRKCB
Clonality: Monoclonal
Clone: A10-F
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR