Anti-PRSS1 Rabbit Monoclonal Antibody [clone: A16-N]

Supplier: Abnova
MAB1641
10685-832EA 346 USD
10685-832
Anti-PRSS1 Rabbit Monoclonal Antibody [clone: A16-N]
Antibodies
Rabbit monoclonal antibody raised against synthetic peptide of PRSS1 .

Recommended Dilutions: Western Blot: 1:4000; ELISA: 1:20000-1:50000

Type: Primary
Antigen: PRSS1
Clonality: Monoclonal
Clone: A16-N
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR