Anti-STRO1 Mouse Monoclonal Antibody [clone: STRO-1]

Supplier: Abnova
MAB10247
10739-970EA 329 USD
10739-970
Anti-STRO1 Mouse Monoclonal Antibody [clone: STRO-1]
Antibodies
Mouse monoclonal antibody raised against of STRO-1.

Type: Primary
Antigen: STRO1
Clonality: Monoclonal
Clone: STRO-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR