Anti-Ig kappa Mouse Monoclonal Antibody [clone: TB28-2] (APC (Allophycocyanin))

Supplier: Abnova
MAB10248
10720-112EA 329 USD
10720-112
Anti-Ig kappa Mouse Monoclonal Antibody [clone: TB28-2] (APC (Allophycocyanin))
Antibodies
Mouse monoclonal antibody raised against of kappa light chains.

Recommended Dilutions: Flow Cytometry: 10 ?l reagent/100 ?l of whole blood or 10^6 cells

Type: Secondary
Antigen: Igkappa
Clonality: Monoclonal
Clone: TB28-2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR