Anti-PDCD4 Mouse Monoclonal Antibody [clone: 9G6]

Supplier: Abnova
MAB10262
10675-096EA 746.62 USD
10675-096
Anti-PDCD4 Mouse Monoclonal Antibody [clone: 9G6]
Antibodies
Mouse monoclonal antibody raised against synthetic phosphopeptide of PDCD4.

Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: PDCD4
Clonality: Monoclonal
Clone: 9G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR