Anti-PTPRC Rabbit Monoclonal Antibody [clone: E19-G]

Supplier: Abnova
MAB1649
10730-716EA 329 USD
10730-716
Anti-PTPRC Rabbit Monoclonal Antibody [clone: E19-G]
Antibodies
Rabbit monoclonal antibody raised against synthetic peptide of PTPRC.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:1000

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: E19-G
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR