Anti-CD163 Rabbit Monoclonal Antibody [clone: K20-T]

Supplier: Abnova
MAB1652
10705-956EA 329 USD
10705-956
Anti-CD163 Rabbit Monoclonal Antibody [clone: K20-T]
Antibodies
Rabbit monoclonal antibody raised against synthetic peptide of CD163.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100

Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: K20-T
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR