Anti-FCAR Mouse Monoclonal Antibody [clone: MIP7c]

Supplier: Abnova
MAB1660
10714-540EA 329 USD
10714-540
Anti-FCAR Mouse Monoclonal Antibody [clone: MIP7c]
Antibodies
Mouse monoclonal antibody raised against partial recombinant FCAR.

Recommended Dilutions: Flow Cytometry: 1 ?g/100 ?l

Type: Primary
Antigen: FCAR
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: MIP7c
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR