Anti-FCAR Mouse Monoclonal Antibody [clone: MIP8a]

Supplier: Abnova
MAB1661
10714-542EA 329 USD
10714-542
Anti-FCAR Mouse Monoclonal Antibody [clone: MIP8a]
Antibodies
Mouse monoclonal antibody raised against partial recombinant FCAR.

Recommended Dilutions: Flow Cytometry: 1 ?g/100 ?l

Type: Primary
Antigen: FCAR
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: MIP8a
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR