Anti-CFP Mouse Monoclonal Antibody [clone: KT20]

Supplier: Abnova
MAB1676
10733-874EA 329 USD
10733-874
Anti-CFP Mouse Monoclonal Antibody [clone: KT20]
Antibodies
Mouse monoclonal antibody raised against native properdin.

Recommended Dilutions: ELISA: 1 ?g/ml

Type: Primary
Antigen: CFP
Clonality: Monoclonal
Clone: KT20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR