Anti-PAX6 Mouse Monoclonal Antibody [clone: 1C8]

Supplier: Abnova
MAB10293
10728-542EA 428.44 USD
10728-542
Anti-PAX6 Mouse Monoclonal Antibody [clone: 1C8]
Antibodies
Mouse monoclonal antibody raised against recombinant PAX6.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: PAX6
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR