Anti-PEG10 Mouse Monoclonal Antibody [clone: 4C10A7]

Supplier: Abnova
MAB10294
10728-952EA 428 USD
10728-952
Anti-PEG10 Mouse Monoclonal Antibody [clone: 4C10A7]
Antibodies
Mouse monoclonal antibody raised against recombinant PEG10.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: PEG10
Clonality: Monoclonal
Clone: 4C10A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR