Anti-RCHY1 Mouse Monoclonal Antibody [clone: 1H10]

Supplier: Abnova
MAB10298
10731-332EA 428.44 USD
10731-332
Anti-RCHY1 Mouse Monoclonal Antibody [clone: 1H10]
Antibodies
Mouse monoclonal antibody raised against recombinant RCHY1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: RCHY1
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR