Anti-PTH1R Mouse Monoclonal Antibody [clone: 4D2]

Supplier: Abnova
MAB10307
10730-592EA 428.44 USD
10730-592
Anti-PTH1R Mouse Monoclonal Antibody [clone: 4D2]
Antibodies
Mouse monoclonal antibody raised against recombinant PTH1R.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: PTH1R
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR