Anti-SKP1 Mouse Monoclonal Antibody [clone: 1H9]

Supplier: Abnova
MAB10320
10737-272EA 428 USD
10737-272
Anti-SKP1 Mouse Monoclonal Antibody [clone: 1H9]
Antibodies
Mouse monoclonal antibody raised against recombinant SKP1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: SKP1
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR