Anti-SORL1 Mouse Monoclonal Antibody [clone: 3B6B11]

Supplier: Abnova
MAB10326
10739-034EA 428.44 USD
10739-034
Anti-SORL1 Mouse Monoclonal Antibody [clone: 3B6B11]
Antibodies
Mouse monoclonal antibody raised against recombinant SORL1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: SORL1
Clonality: Monoclonal
Clone: 3B6B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR