Anti-FFAR4 Mouse Monoclonal Antibody [clone: 2B6]

Supplier: Abnova
MAB1725
10682-564EA 747 USD
10682-564
Anti-FFAR4 Mouse Monoclonal Antibody [clone: 2B6]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of O3FAR1.

Recommended Dilutions: Immunoprecipitation: 5 ?g/ml; Flow Cytometry: 1-5 ?g/ml

Type: Primary
Antigen: O3FAR1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 2B6
Epitope:
Host: IgG2a kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR