Anti-VTG Mouse Monoclonal Antibody [clone: 3E11]

Supplier: Abnova
MAB1728
10744-356EA 531 USD
10744-356
Anti-VTG Mouse Monoclonal Antibody [clone: 3E11]
Antibodies
Mouse monoclonal antibody raised against native vitellogenin (VTG).

Recommended Dilutions: Western Blot: 1 ?g/ml; ELISA: 1-2 ?g/ml

Type: Primary
Antigen: VTG
Clonality: Monoclonal
Clone: 3E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR