Anti-TNFRSF11B Mouse Monoclonal Antibody [clone: 5G2]

Supplier: Abnova
MAB10335
10742-068EA 428.44 USD
10742-068
Anti-TNFRSF11B Mouse Monoclonal Antibody [clone: 5G2]
Antibodies
Mouse monoclonal antibody raised against recombinant TNFRSF11B.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: TNFRSF11B
Clonality: Monoclonal
Clone: 5G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR