Anti-PAK2 Mouse Monoclonal Antibody [clone: 3B5]

Supplier: Abnova
MAB10355
10733-328EA 530.32 USD
10733-328
Anti-PAK2 Mouse Monoclonal Antibody [clone: 3B5]
Antibodies
Mouse monoclonal antibody raised against recombinant PAK2.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: PAK2
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR