Anti-PROZ Mouse Monoclonal Antibody [clone: 2B4]

Supplier: Abnova
MAB10357
10730-282EA 428.44 USD
10730-282
Anti-PROZ Mouse Monoclonal Antibody [clone: 2B4]
Antibodies
Mouse monoclonal antibody raised against recombinant PROZ.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: PROZ
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR